Learn More
Strem, An Ascensus Company CAS# 18899-64-4. 500mg. 1,2-Bis(phenylphoshino)ethane, min. 90%. MFCD00003041
SDP
Supplier:  Strem, An Ascensus Company 150405500MG
Encompass
CAS# 18899-64-4. 500mg. 1,2-Bis(phenylphoshino)ethane, min. 90%. MFCD00003041. Molecular Weight: 246.22. Molecular Formula: (C6H5)HPCH2CH2PH(C6H5). Color/form: colorless to yellow liq. Strem# 15-0405. http://www.strem.com/catalog/v/15-0405/
Catalog No. 50-368-2