Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(trichlorosilyl)ethane, Thermo Scientific Chemicals

Catalog No. AC462030250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462030250 25 g
AC462030050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462030250 Supplier Thermo Scientific Chemicals Supplier No. 462030250
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2504-64-5
Molecular Formula C2H4Cl6Si2
Molecular Weight (g/mol) 296.92
InChI Key WDVUXWDZTPZIIE-UHFFFAOYSA-N
IUPAC Name trichloro[2-(trichlorosilyl)ethyl]silane
SMILES Cl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl

Specifications

Melting Point 24.0°C to 25.0°C
Flash Point 65°C
Quantity 25 g
Formula Weight 296.94
Chemical Name or Material 1,2-Bis(trichlorosilyl)ethane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.

Reacts violently with water.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 219-710-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.