Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Bis(triethoxysilyl)ethane

Catalog No. 44725025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-725-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-725-025ML Supplier Sigma Aldrich Supplier No. 44725025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-bis(triethoxysilyl)ethane is a non-functional silane and a double trialkoxysilyl precursor.BTESE can be used as an anti-corrosive coating on aluminium magnesium alloy (Al93Mg7) for potential application in nuclear and food processing industries. BTESE membranes are chlorine resistant and can be thermally stable at 100°C. They which can be used in gas permeation (GS) and reverse osmosis (RO) applications. It may also form a mesoporous hybrid monolith with tetramethoxysilane by a sol-gel condensation method with enhanced chemical stability for peptide separation.

Specifications

CAS 16068-37-4
Density 0.958 g/mL (at 25°C)
Boiling Point 119°C
Molecular Formula C14H34O6Si2
Refractive Index n20/D 1.411 (literature)
Linear Formula [-CH2Si(OC2H5)3]2
Quantity 25 mL
MDL Number MFCD00239362
Synonym 1,2-(Triethoxysilyl)ethane; 1,2-Bis(triethoxysilyl)ethane; 4,4,7,7-Tetraethoxy-3,8-dioxa-4,7-disiladecane; Ethylenebis(triethoxysilane)
Molecular Weight (g/mol) 354.59
Percent Purity 96%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.