Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(triethoxysilyl)ethane, 95%

Catalog No. AAH60530AC
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60530AC 25 mL
AAH60530AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60530AC Supplier Thermo Scientific Chemicals Supplier No. H60530AC
  • $120.40
    $96.65 / Each of 1

    Save $23.75

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16068-37-4 | C14H34O6Si2 | 354.59 g/mol

1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(triethoxysilyl)ethane is used as coating auxiliary agents, leather auxiliary agents, plastic auxiliary agents, surfactants and textile auxiliary agents. It is used to prepare mesoporous organosilica materials.

Solubility
Soluble in alcohol, aromatic and aliphatic hydrocarbons. Hydrolyzes with water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16068-37-4
Molecular Formula C14H34O6Si2
Molecular Weight (g/mol) 354.59
MDL Number MFCD00239362
InChI Key IZRJPHXTEXTLHY-UHFFFAOYSA-N
Synonym 1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane
PubChem CID 85266
IUPAC Name 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane
SMILES CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC

Specifications

Boiling Point 119°C
Quantity 25 mL
Flash Point 113°C (235°F)
Refractive Index 1.411
Density 0.958
Linear Formula [-CH2Si(OC2H5)3]2
Solubility Information Soluble in alcohol,aromatic and aliphatic hydrocarbons. Hydrolyzes with water.
Formula Weight 354.59
Percent Purity 95%
Chemical Name or Material 1,2-Bis(triethoxysilyl)ethane
Hazard Category H301-H312-H372
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P312-P314-P330-P363-P501c
EINECSNumber 240-212-2
RTECSNumber JG7755000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.