Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(triethoxysilyl)ethane 95.0+%, TCI America™
SDP

Catalog No. B41235ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41235ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41235ML Supplier TCI America Supplier No. B41235ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16068-37-4
Molecular Formula C14H34O6Si2
Molecular Weight (g/mol) 354.59
MDL Number MFCD00239362
InChI Key IZRJPHXTEXTLHY-UHFFFAOYSA-N
Synonym 1,2-bis triethoxysilyl ethane, 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane, 3,8-dioxa-4,7-disiladecane, 4,4,7,7-tetraethoxy, ethylenebis triethoxysilane, bis triethoxysilyl ethane, hexaethoxydisilethylene, bis 1,2-triethoxysilyl ethane, 1,2-bis triethoxysilyl ethane,, triethoxysilyl ethyl triethoxysilane, triethoxy 2-triethoxysilylethyl silane
PubChem CID 85266
IUPAC Name 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane
SMILES CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 mL
Formula Weight 354.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(triethoxysilyl)ethane
EINECSNumber (2)-3975
RTECSNumber JG7755000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.