Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(trimethylsiloxy)ethane, 98%

Catalog No. AAH6385709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6385709 10 g
AAH6385718 50 g
AAH6385730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6385709 Supplier Thermo Scientific Chemicals Supplier No. H6385709
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 7381-30-8 | C8H22O2Si2 | 206.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7381-30-8
Molecular Formula C8H22O2Si2
Molecular Weight (g/mol) 206.43
MDL Number MFCD00009640
InChI Key JGWFUSVYECJQDT-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsiloxy ethane, 1,2-bis trimethylsilyloxy ethane, 2,2,7,7-tetramethyl-3,6-dioxa-2,7-disilaoctane, ethylene glycol bis trimethylsilyl ether, ethylenedioxybis trimethylsilane, 3,6-dioxa-2,7-disilaoctane, 2,2,7,7-tetramethyl, 1,2-bis-trimethylsilyloxy ethane, 1,2-bis-trimethylsilanyloxy-ethane, ethylene glycol, di-tms, acmc-1bh50
PubChem CID 81858
SMILES C[Si](C)(C)OCCO[Si](C)(C)C

Specifications

Boiling Point 165°C to 171°C
Quantity 10 g
Linear Formula (CH3)3SiOCH2CH2OSi(CH3)3
UN Number UN1993
Beilstein 1744217
Flash Point 51°C (124°F)
IUPAC Name trimethyl(2-trimethylsilyloxyethoxy)silane
Formula Weight 206.43
Percent Purity 98%
Refractive Index 1.403
Sensitivity Moisture sensitive
Density 0.842
Chemical Name or Material 1,2-Bis(trimethylsiloxy)ethane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 230-950-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.