Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(trimethylsilyl)benzene 95.0+%, TCI America™
SDP

Catalog No. B22995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22995G 5 g
B22991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22995G Supplier TCI America Supplier No. B22995G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17151-09-6
Molecular Formula C12H22Si2
Molecular Weight (g/mol) 222.478
MDL Number MFCD00015590
InChI Key YHMJZIJXVNRXIN-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsilyl benzene, trimethyl 2-trimethylsilyl phenyl silane, trimethyl-2-trimethylsilylphenyl silane, acmc-1c8d9, benzene,1,2-bis trimethylsilyl, 1,2-phenylenebis trimethylsilane, trimethyl 2-trimethylsilyl phenyl silane #
PubChem CID 519794
IUPAC Name trimethyl-(2-trimethylsilylphenyl)silane
SMILES C[Si](C)(C)C1=CC=CC=C1[Si](C)(C)C

Specifications

Color Colorless
Quantity 5 g
Formula Weight 222.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trimethylsilyl)benzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.