Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethyl)-3-(trifluoromethyl)benzene, 95%, Thermo Scientific™

Catalog No. p-7055894
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6460103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6460103 Supplier Thermo Scientific Chemicals Supplier No. H6460103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1997-80-4
Molecular Formula C9H8BrF3
Molecular Weight (g/mol) 253.062
MDL Number MFCD03092900
InChI Key AZLYAPKPZBXCGX-UHFFFAOYSA-N
Synonym 1-2-bromoethyl-3-trifluoromethyl benzene, 3-trifluoromethyl phenethyl bromide, 3-trifluoromethyl phenylethyl bromide, benzene, 1-2-bromoethyl-3-trifluoromethyl, 1-bromo-2-3-trifluoromethyl phenyl ethane, acmc-20anht, 3-trifluoromethylphenethyl bromide, toluene, m-2-bromoethyl-.alpha.,.alpha.,.alpha.-trifluoro, 3-trifluoromethyl phenylethylbromide
PubChem CID 594535
IUPAC Name 1-(2-bromoethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCBr

Specifications

Color Colorless
Density 1.495 g/mL
Boiling Point 92°C to 94°C (12 mmHg)
Refractive Index 1.486
Quantity 1 g
UN Number UN3082
Beilstein 1950052
Formula Weight 253.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)-3-(trifluoromethyl)benzene
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.