Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B52451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52451G 1 g
B52455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52451G Supplier TCI America Supplier No. B52451G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1997-80-4
Molecular Formula C9H8BrF3
Molecular Weight (g/mol) 253.062
MDL Number MFCD03092900
InChI Key AZLYAPKPZBXCGX-UHFFFAOYSA-N
Synonym 1-2-bromoethyl-3-trifluoromethyl benzene, 3-trifluoromethyl phenethyl bromide, 3-trifluoromethyl phenylethyl bromide, benzene, 1-2-bromoethyl-3-trifluoromethyl, 1-bromo-2-3-trifluoromethyl phenyl ethane, acmc-20anht, 3-trifluoromethylphenethyl bromide, toluene, m-2-bromoethyl-.alpha.,.alpha.,.alpha.-trifluoro, 3-trifluoromethyl phenylethylbromide
PubChem CID 594535
IUPAC Name 1-(2-bromoethyl)-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCBr

Specifications

Color Yellow
Boiling Point 94°C
Quantity 1 g
Formula Weight 253.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Bromoethyl)-3-(trifluoromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.