Learn More
Medchemexpress LLC 1-(2-bromophenyl)-3-(7-cyano-2H-benzotriazol-4-yl)urea | 211096-49-0 | MFCD09971124 | 699.0% | 357.16 g·mol⁻¹ | C14H9BrN6O | 10 MG
SDP
Catalog No. 5000298498
Encompass_Preferred
SB-265610 is a selective, competitive, nonpeptide, allosteric antagonist of the CXCR2 chemokine receptor used as a research reagent to study neutrophil chemotaxis and CXCR2-mediated signaling. It inhibits CINC-1-induced calcium mobilization (IC50 3.7 nM) and neutrophil chemotaxis (IC50 70 nM).
  • Selective CXCR2 antagonist for receptor pharmacology studies.
  • Reported low-nanomolar potency for calcium mobilization assays.
  • Suitable for in vitro chemotaxis and signaling experiments.
  • Supplied as a small-molecule solid for formulation or dosing control.
  • Characterized by published analytical data and CAS registration for traceability.
Catalog No. 50-002-98498 Supplier Medchemexpress LLC Supplier No. HY5068810MG
SB-265610 is a selective, competitive, nonpeptide, allosteric antagonist of the CXCR2 chemokine receptor used as a research reagent to study neutrophil chemotaxis and CXCR2-mediated signaling. It inhibits CINC-1-induced calcium mobilization (IC50 3.7 nM) and neutrophil chemotaxis (IC50 70 nM).
  • Selective CXCR2 antagonist for receptor pharmacology studies.
  • Reported low-nanomolar potency for calcium mobilization assays.
  • Suitable for in vitro chemotaxis and signaling experiments.
  • Supplied as a small-molecule solid for formulation or dosing control.
  • Characterized by published analytical data and CAS registration for traceability.