Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Butanediol

Catalog No. 189301L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-930-1L 1 L
18-930-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-930-1L Supplier Sigma Aldrich Supplier No. 189301L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,2-butanediol has been used:
  • To investigate dilatometric measurements of excess molar volumes and excess partial molar volumes for binary mixtures of n,n-dimethylformamide with 1,2-alkanediols
  • In the preparation of 1,2-diolates of titanium by reaction with titanium (iv) isopropoxide or titanium (iv) chloride

Specifications

CAS 584-03-2
Density 1.006 g/mL (at 25°C (literature))
Boiling Point 191°C to 192°C (747 mmHg)
Molecular Formula C4H10O2
Linear Formula CH3CH2CH(OH)CH2OH
Refractive Index n20/D 1.438 (literature); n20/D 1.438
MDL Number MFCD00004570
Quantity 1 L
Synonym 1,2-Butylene glycol
Molecular Weight (g/mol) 90.12
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.