Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Butanediol, 98%, Thermo Scientific™

Catalog No. AC175990010
Change view
Click to view available options
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC175990010 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175990010 Supplier Thermo Scientific Chemicals Supplier No. 175990010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 584-03-2
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C4H10O2
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
Linear Formula C2H5CH(OH)CH2OH
Quantity 1L
MDL Number MFCD00004570
Beilstein 01, 477
Synonym 1,2-butanediol, 1,2-butylene glycol, 1,2-dihydroxybutane, alpha-butyleneglycol, alpha-butylene glycol, 1,2-butandiol, 1,2-dihydroxy butane, butanediol, 1,2, .alpha.-butylene glycol, 1,2-butylene-glycol
InChI Key BMRWNKZVCUKKSR-UHFFFAOYNA-N
SMILES CCC(O)CO
IUPAC Name butane-1,2-diol
Molecular Weight (g/mol) 90.12
PubChem CID 11429
ChEBI CHEBI:52682
Formula Weight 90.12
Percent Purity ≥97.5%
Chemical Name or Material 1, 2-Butanediol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.