Learn More
World Precision Instrument #1/2 Carbide Ball Mill
SDP
Catalog No. NC1689491
Encompass
This line-powered micro drill will make easy work of grinding, finishing, cutting and drilling bone, teeth and other material. The high-torque 35,000 rpm (maximum) motor is quiet and has minimal vibration which reduces wear on the motor and provides greater comfort for you. It also features a forward and reverse switch, “E Type” handpiece and handpiece holder. The handpiece has a removable nose cone that can be cleaned and sterilized. It accepts 3/32” and 2.33mm bur shanks. Unlike battery-powered drills, this unit maintains consistent power for the duration of use. The wide range of speeds allows you to control the amount of heat generation.
Catalog No. NC1689491 Supplier World Precision Instrument Supplier No. 501861
This line-powered micro drill will make easy work of grinding, finishing, cutting and drilling bone, teeth and other material. The high-torque 35,000 rpm (maximum) motor is quiet and has minimal vibration which reduces wear on the motor and provides greater comfort for you. It also features a forward and reverse switch, “E Type” handpiece and handpiece holder. The handpiece has a removable nose cone that can be cleaned and sterilized. It accepts 3/32” and 2.33mm bur shanks. Unlike battery-powered drills, this unit maintains consistent power for the duration of use. The wide range of speeds allows you to control the amount of heat generation.