Learn More
Medchemexpress LLC 1-(2-chloroethyl)-3-((1R,4R)-4-hydroxycyclohexyl)-1-nitrosourea-d4 | 00-00-0 | 99.0% | 253.72 | C9H12D4ClN3O3 | 5 MG
SDP
Catalog No. 5000222654
Encompass_Preferred
Deuterium-labeled trans-4'-Hydroxy CCNU Lomustine-d4 is a research-grade labeled analogue of trans-4'-Hydroxy lomustine intended for use as a tracer and internal standard in quantitative analytical methods. The product is supplied as a high-purity solid in a 5 MG vial and requires cold storage to maintain stability.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High purity (99.0%).
  • Solid, white to off-white appearance.
  • Suitable as a tracer for pharmacokinetic and metabolic studies.
  • Powder storage: -20°C for long-term stability; in solvent: -80°C for short-term stability.
  • Supplied as a 5 MG vial for laboratory use.
Catalog No. 50-002-22654 Supplier Medchemexpress LLC Supplier No. HY143979S5MG
Deuterium-labeled trans-4'-Hydroxy CCNU Lomustine-d4 is a research-grade labeled analogue of trans-4'-Hydroxy lomustine intended for use as a tracer and internal standard in quantitative analytical methods. The product is supplied as a high-purity solid in a 5 MG vial and requires cold storage to maintain stability.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High purity (99.0%).
  • Solid, white to off-white appearance.
  • Suitable as a tracer for pharmacokinetic and metabolic studies.
  • Powder storage: -20°C for long-term stability; in solvent: -80°C for short-term stability.
  • Supplied as a 5 MG vial for laboratory use.