Learn More
Medchemexpress LLC 1-(2-Chloroethyl)-3-((1R,4R)-4-hydroxycyclohexyl)-1-nitrosourea-d4 | 56239-24-8 | 99.0% | C9H12D4ClN3O3 | 1 MG
SDP
Catalog No. 5000355651
Encompass_Preferred
trans-4'-Hydroxy CCNU Lomustine-d4 is a deuterium-labeled compound, primarily used as a tracer or as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, like deuterium, into drug molecules serves as a critical method for quantitation during drug development, with the potential to influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-55651 Supplier Medchemexpress LLC Supplier No. HY143979S1MG
trans-4'-Hydroxy CCNU Lomustine-d4 is a deuterium-labeled compound, primarily used as a tracer or as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, like deuterium, into drug molecules serves as a critical method for quantitation during drug development, with the potential to influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs.