Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Chloroethyl)naphthalene 95.0+%, TCI America™
SDP

Catalog No. C14905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14905G 5 g
C149025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14905G Supplier TCI America Supplier No. C14905G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41332-02-9
Molecular Formula C12H11Cl
Molecular Weight (g/mol) 190.67
MDL Number MFCD01321149
InChI Key COIBFCUQUWJKEN-UHFFFAOYSA-N
PubChem CID 3540711
IUPAC Name 1-(2-chloroethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCCl

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 190.67
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Chloroethyl)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.