Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Chloroethyl)-pyrrolidine

Catalog No. CDS008148 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS008148 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS008148 Supplier Sigma Aldrich Supplier No. CDS008148250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Direct asymmetric reaction of dibenzyl malonate with n-tert-butoxycarbonyl aldimines in the presence of yb(otf)3 and ipr-pybox (pybox = pyridine bisoxazoline) complexes has been investigated.dibenzyl malonate was used in the preparation of tetraethyl 3,3-bis(benzyloxycarbonyl)propylene bisphosphonate
  • It was also used in the preparation of benzyl umbelliferone-3-carboxylate via knoevenagel condensation with 2,4-dihydroxybezaldehyde

Specifications

CAS 5050-41-9
Molecular Formula C6H12ClN
Linear Formula C6H12ClN
MDL Number MFCD00044568
Quantity 50 g
Molecular Weight (g/mol) 133.62
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.