Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Chlorophenyl)-3-methyl-5-pyrazolone 98.0+%, TCI America™
SDP

Catalog No. C341425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C341425G 25 g
C34145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C341425G Supplier TCI America Supplier No. C341425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14580-22-4
Molecular Formula C10H9ClN2O
Molecular Weight (g/mol) 208.645
MDL Number MFCD00059717
InChI Key CWESERWNUIUBJU-UHFFFAOYSA-N
PubChem CID 84534
IUPAC Name 2-(2-chlorophenyl)-5-methyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C2=CC=CC=C2Cl

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 208.65
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 1-(2-Chlorophenyl)-3-methyl-5-pyrazolone
EINECSNumber (5)-0302
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.