Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Chlorophenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. C11025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11025G 5 g
C110225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11025G Supplier TCI America Supplier No. C11025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39512-50-0
Molecular Formula C10H13ClN2
Molecular Weight (g/mol) 196.678
MDL Number MFCD00040727
InChI Key PWZDJIUQHUGFRJ-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl piperazine, 1-2-chloro-phenyl-piperazine, piperazine, 1-2-chlorophenyl, chembl9841, 1-o-chlorophenyl piperazine, 2-chlorophenyl piperazine, chlorphenylpiperazin, pubchem14571, acmc-1aif1
PubChem CID 415628
IUPAC Name 1-(2-chlorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=CC=C2Cl

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 196.68
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(2-Chlorophenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.