Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Cyanoethyl)piperidine-3-carboxylic acid

Catalog No. OTV000428 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000428 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000428 Supplier Sigma Aldrich Supplier No. OTV000428250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • (+/-)-3-benzyloxy-1,2-propanediol undergoes enantioseparation by ligand exchange micellar electrokinetic chromatography using borate anion as a central ion.(+/-)-3-benzyloxy-1,2-propanediol was used in capillary electrophoretic enantioseparation of vicinol diols using different α-cyclodextrin derivatives and borate
  • It was used in the synthesis and immobilization of 2,3-di-o-phytanyl-sn-glycerol-1-tetraethylene glycol-(3-trichloropropyl-silane) ether lipid (dpttc) and 2,3-di-o-phytanyl-sn-glycerol-1-tetraethylene glycol-(3-chloro-dimethylpropyl-silane) ether lipid(dptdc)

Specifications

Molecular Formula C9H14N2O2
Linear Formula C9H14N2O2
Quantity 10 mL
MDL Number MFCD11914319
Molecular Weight (g/mol) 182.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.