Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)-5-iodouracil 98.0+%, TCI America™
SDP

Catalog No. D420010MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D420010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D420010MG Supplier TCI America Supplier No. D420010MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69123-98-4
Molecular Formula C9H10FIN2O5
Molecular Weight (g/mol) 372.091
MDL Number MFCD00866922
InChI Key IPVFGAYTKQKGBM-BYPJNBLXSA-N
Synonym Fialuridine, FIAU, 2′C-Fluoro-5-iodoarauracil
PubChem CID 50313
IUPAC Name 1-[(2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)F)I

Specifications

Melting Point 226°C
Color White
Quantity 10 mg
Formula Weight 372.09
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)-5-iodouracil
RTECSNumber YQ9512500
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.