Learn More
Medchemexpress LLC 1-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil | 69123-94-0 | MFCD00870798 | ≥98.0% | 246.19 | C9H11FN2O5 | 1 G
SDP
Catalog No. 5000301658
Encompass_Preferred
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil (CAS 69123-94-0) is a fluorinated deoxy-arabinofuranosyl uracil nucleoside analogue with reported antiviral activity. It is provided for research use and is characterized by molecular formula C9H11FN2O5 and molecular weight 246.19 g·mol-1.
  • Fluorinated nucleoside analogue suitable for antiviral mechanism studies.
  • High purity (≥98.0%) for reproducible experimental results.
  • Available in solid form and as a DMSO solution for flexible use.
  • Well-characterized chemical properties for analytical verification.
  • Intended for research use only, not for human or veterinary use.
Catalog No. 50-003-01658 Supplier Medchemexpress LLC Supplier No. HYW0177491G
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil (CAS 69123-94-0) is a fluorinated deoxy-arabinofuranosyl uracil nucleoside analogue with reported antiviral activity. It is provided for research use and is characterized by molecular formula C9H11FN2O5 and molecular weight 246.19 g·mol-1.
  • Fluorinated nucleoside analogue suitable for antiviral mechanism studies.
  • High purity (≥98.0%) for reproducible experimental results.
  • Available in solid form and as a DMSO solution for flexible use.
  • Well-characterized chemical properties for analytical verification.
  • Intended for research use only, not for human or veterinary use.