Learn More
Medchemexpress LLC 1-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil | 69123-94-0 | MFCD00870798 | ≥98.0% | 246.19 | C9H11FN2O5 | 1 ML
SDP
Catalog No. 5000300648
Encompass_Preferred
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil is a fluorinated nucleoside analogue with reported antiviral activity, supplied for research use. It is available as a solid or as a 10 mM solution in DMSO, and is provided with supporting documentation for quality control. The compound has molecular formula C9H11FN2O5, molecular weight 246.19 g/mol, and a typical purity of ≥98.0%.
  • Reported antiviral activity relevant to antiviral and nucleoside analogue research.
  • Available as a solid or a ready-to-use 10 mM solution in DMSO for convenience.
  • High purity suitable for analytical, biochemical, and cellular assays.
  • Well-characterized chemical properties for formulation and experimental planning.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for traceability.
Catalog No. 50-003-00648 Supplier Medchemexpress LLC Supplier No. HYW01774910MM/1ML
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil is a fluorinated nucleoside analogue with reported antiviral activity, supplied for research use. It is available as a solid or as a 10 mM solution in DMSO, and is provided with supporting documentation for quality control. The compound has molecular formula C9H11FN2O5, molecular weight 246.19 g/mol, and a typical purity of ≥98.0%.
  • Reported antiviral activity relevant to antiviral and nucleoside analogue research.
  • Available as a solid or a ready-to-use 10 mM solution in DMSO for convenience.
  • High purity suitable for analytical, biochemical, and cellular assays.
  • Well-characterized chemical properties for formulation and experimental planning.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for traceability.