Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2-deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil | 69123-94-0 | MFCD00870798 | 98.0% | 246.19 g·mol⁻1 | C9H11FN2O5 | 500 MG
SDP

Catalog No. 5000301659
Encompass_Preferred

1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil is a fluorinated nucleoside analogue provided for research use. It exhibits reported antiviral activity and is characterized by defined physicochemical properties and batch-specific quality documentation for reliable use in in vitro and in vivo studies.

  • Fluorinated nucleoside analogue with reported antiviral activity.
  • Purity ≥98.0%.
  • Chemical formula C9H11FN2O5; molecular weight 246.19 g·mol⁻1.
  • White to off-white solid form, soluble in DMSO (100 mg/mL).
  • Storage stability: powder -20°C (long-term) or 4°C (short-term); solutions -80°C for extended storage.
  • Supplied with batch-specific certificate of analysis for quality verification.

Catalog No. 50-003-01659 Supplier Medchemexpress LLC Supplier No. HYW017749500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)uracil is a fluorinated nucleoside analogue provided for research use. It exhibits reported antiviral activity and is characterized by defined physicochemical properties and batch-specific quality documentation for reliable use in in vitro and in vivo studies.

  • Fluorinated nucleoside analogue with reported antiviral activity.
  • Purity ≥98.0%.
  • Chemical formula C9H11FN2O5; molecular weight 246.19 g·mol⁻1.
  • White to off-white solid form, soluble in DMSO (100 mg/mL).
  • Storage stability: powder -20°C (long-term) or 4°C (short-term); solutions -80°C for extended storage.
  • Supplied with batch-specific certificate of analysis for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.