Learn More
Medchemexpress LLC 1-(2-Deoxy-β-D-threo-pentofuranosyl)thymine | 16053-52-4 | C10H14N2O5 | 25 MG
SDP
Catalog No. 5000375043
Encompass_Preferred
1-(2-Deoxy-β-D-threo-pentofuranosyl)thymine is a thymidine analog with a purity of 99.68% and a molecular weight of 242.23. This compound exhibits insertional activity towards replicated DNA, making it suitable for labeling cells and tracking DNA synthesis. It also demonstrates antiviral activity against HBV in human 2.2.15 cells with an EC50 of 10 μg/mL and against DHBV in duck hepatocytes with an EC50 of 4.13 μM. Cytotoxicity against human HuH7 cells is observed with a CC50 of > 200 μg/mL.
  • Thymidine analog
  • Exhibits insertional activity towards replicated DNA
  • Used for labeling cells and tracking DNA synthesis
  • Demonstrates antiviral activity against HBV and DHBV
  • High purity of 99.68%
  • Appears as a white to off-white solid
Catalog No. 50-003-75043 Supplier Medchemexpress LLC Supplier No. 25MGHY154524
1-(2-Deoxy-β-D-threo-pentofuranosyl)thymine is a thymidine analog with a purity of 99.68% and a molecular weight of 242.23. This compound exhibits insertional activity towards replicated DNA, making it suitable for labeling cells and tracking DNA synthesis. It also demonstrates antiviral activity against HBV in human 2.2.15 cells with an EC50 of 10 μg/mL and against DHBV in duck hepatocytes with an EC50 of 4.13 μM. Cytotoxicity against human HuH7 cells is observed with a CC50 of > 200 μg/mL.
  • Thymidine analog
  • Exhibits insertional activity towards replicated DNA
  • Used for labeling cells and tracking DNA synthesis
  • Demonstrates antiviral activity against HBV and DHBV
  • High purity of 99.68%
  • Appears as a white to off-white solid