Learn More
Medchemexpress LLC 1-(2-Deoxy-β-D-threo-pentofuranosyl)thymine | 16053-52-4 | C10H14N2O5 | 50 MG
SDP
Catalog No. 5000376274
Encompass_Preferred
This product, 1-(2-Deoxy-β-D-threo-pentofuranosyl)thymine, is a laboratory chemical primarily used in research. It functions as a thymidine analog, exhibiting insertional activity towards replicated DNA. It is supplied as a solid.
  • Thymidine analog
  • Exhibits insertional activity towards replicated DNA
  • Suitable for laboratory chemical applications
  • Supplied as a solid
  • Melting point 172-173°C
Catalog No. 50-003-76274 Supplier Medchemexpress LLC Supplier No. HY15452450MG
This product, 1-(2-Deoxy-β-D-threo-pentofuranosyl)thymine, is a laboratory chemical primarily used in research. It functions as a thymidine analog, exhibiting insertional activity towards replicated DNA. It is supplied as a solid.
  • Thymidine analog
  • Exhibits insertional activity towards replicated DNA
  • Suitable for laboratory chemical applications
  • Supplied as a solid
  • Melting point 172-173°C