Learn More
Medchemexpress LLC 1,2-di-o-hexadecyl-sn-glycero-3-phosphocholine | 36314-47-3 | 97.0% | 706.07 g/mol | C40H84NO6P | 5 MG
SDP
Catalog No. 5000262937
Encompass_Preferred
1,2-Di-O-hexadecyl-sn-glycero-3-phosphocholine is a synthetic ether-linked phospholipid bearing hexadecyl (C16) ether chains at the sn-1 and sn-2 positions of glycerol. It is commonly used to prepare stable liposomes and model membranes for membrane biophysics and formulation studies, offering greater resistance to hydrolysis than ester-linked phospholipids.
  • Stable ether-linked phospholipid suitable for liposome formation.
  • Resistant to hydrolysis, improving membrane stability.
  • High purity appropriate for research applications.
  • Solid form simplifies storage and handling.
Catalog No. 50-002-62937 Supplier Medchemexpress LLC Supplier No. HY1316525MG
1,2-Di-O-hexadecyl-sn-glycero-3-phosphocholine is a synthetic ether-linked phospholipid bearing hexadecyl (C16) ether chains at the sn-1 and sn-2 positions of glycerol. It is commonly used to prepare stable liposomes and model membranes for membrane biophysics and formulation studies, offering greater resistance to hydrolysis than ester-linked phospholipids.
  • Stable ether-linked phospholipid suitable for liposome formation.
  • Resistant to hydrolysis, improving membrane stability.
  • High purity appropriate for research applications.
  • Solid form simplifies storage and handling.