Learn More
Medchemexpress LLC 1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine | 36314-47-3 | >98.0% | C40H84NO6P | 10MG
SDP
Catalog No. 5000260961
Encompass_Preferred
1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine (CAS 36314-47-3) is an ether-linked phosphatidylcholine used as a synthetic, nonhydrolysable phospholipid analogue for membrane and liposome research. It is applied in biophysical studies and drug delivery research to create stable model membranes and liposomal formulations.
  • Ether-linked phospholipid resistant to enzymatic hydrolysis.
  • Supports formation of stable liposomes and model membranes.
  • Useful in membrane biophysics and drug delivery studies.
  • Hydrophobic alkyl chains provide low permeability and increased membrane rigidity.
  • Supplied as a powder for solvent-based formulation and dispersion.
Catalog No. 50-002-60961 Supplier Medchemexpress LLC Supplier No. HY13165210MG
1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine (CAS 36314-47-3) is an ether-linked phosphatidylcholine used as a synthetic, nonhydrolysable phospholipid analogue for membrane and liposome research. It is applied in biophysical studies and drug delivery research to create stable model membranes and liposomal formulations.
  • Ether-linked phospholipid resistant to enzymatic hydrolysis.
  • Supports formation of stable liposomes and model membranes.
  • Useful in membrane biophysics and drug delivery studies.
  • Hydrophobic alkyl chains provide low permeability and increased membrane rigidity.
  • Supplied as a powder for solvent-based formulation and dispersion.