Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole 98.0+%, TCI America™
SDP

Catalog No. D38345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38345G 5 g
D38341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38345G Supplier TCI America Supplier No. D38345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 628333-86-8
Molecular Formula C29H33N2P
Molecular Weight (g/mol) 440.571
MDL Number MFCD09038823
InChI Key JAIIBHBCNPAPPF-UHFFFAOYSA-N
Synonym TrippyPhos
PubChem CID 11430600
IUPAC Name ditert-butyl-[2-(3,5-diphenylpyrazol-1-yl)phenyl]phosphane
SMILES CC(C)(C)P(C1=CC=CC=C1N2C(=CC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C(C)(C)C

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 440.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.