Learn More
Electron Microscopy Sciences EMS 1/2" dia Aperture Flamer
SDP
Catalog No. 50980726
Encompass_Preferred
For holding aperture discs or strips while flaming. Small discs can be flamed in the 3/16" diameter cup made from 6 mil platinum foil with a 1/32" diameter hole in the bottom to induce a good flow of air. The cup is attached by a 30 mil platinum wire to a heat resistant handle. The aperture can be hung from the hook for flaming.
For holding 1/4" to 1/2" diameter aperture discs while flaming. The flat pan is made from 6 mil platinum foil with a 1/32" diameter hole in the bottom to induce adequate air flow. A 30 mil platinum wire attaches the pan to a heat resistant handle.
Catalog No. 50-980-726 Supplier Electron Microscopy Sciences Supplier No. 40002
For holding aperture discs or strips while flaming. Small discs can be flamed in the 3/16" diameter cup made from 6 mil platinum foil with a 1/32" diameter hole in the bottom to induce a good flow of air. The cup is attached by a 30 mil platinum wire to a heat resistant handle. The aperture can be hung from the hook for flaming.
For holding 1/4" to 1/2" diameter aperture discs while flaming. The flat pan is made from 6 mil platinum foil with a 1/32" diameter hole in the bottom to induce adequate air flow. A 30 mil platinum wire attaches the pan to a heat resistant handle.