Learn More
Sigma Aldrich 1,2-diamino-3,5-difluorobenzene
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Methyl 10-undecenoate is the chain stopper in the polymerization of long-chain aliphatic α,-d-diene through acyclic diene metathesis (admet). this helps in tuning the molecular weight of the resulting polyester in a range varying from 10 to 45 kda.methyl 10-undecenoate is an unsaturated fatty acid ester that can be used as a substrate:
- To prepare bifunctional polymer precursors via hydroformylation reaction
- To synthesize α,-d-ester amides via aminocarbonylation in the presence of palladium catalyst
- To modify commercial silicone monomer, 1,3,5,7-tetramethylcyclotetrasiloxane, for the preparation of silicone hollow nano/microstructures in water by hydrosilylation reaction
Specifications
Specifications
| Molecular Formula | C6H6F2N2 |
| Linear Formula | C6H6F2N2 |
| MDL Number | MFCD00973899 |
| Quantity | 500 mL |
| Molecular Weight (g/mol) | 144.12 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.