Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminocyclohexanetetraacetic acid disodium salt solution, Solstice

Catalog No. 6002597 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-97 1 L
60-046-531 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-025-97 Supplier Solstice Supplier No. 351031L
Add to Cart
Add to Cart

Volumetric, Reag. Ph. Eur., 0.1 M DCTA-Na4, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5786-78-7
Molecular Formula C14H20N2Na2O8
Molecular Weight (g/mol) 390.3
InChI Key TVTLKUMLAQUFAS-UHFFFAOYSA-L
Synonym sodium 1,2-diaminocyclohexane tetraacetate, 1,2-diaminocyclohexenetetra-acetate of sodium, acetic acid, 1,2-cyclohexylenedinitrilo tetra-, sodium salt, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, disodium salt, sodium n'-1,2-cyclohexanediylbis n-carboxymethyl glycinate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, sodium salt 1:2, cdta disodium salt, disodium 2-carboxymethyl-2-carboxymethyl-2-oxido-2-oxoethyl amino cyclohexyl amino acetate
PubChem CID 24230
IUPAC Name disodium;2-[[2-[carboxylatomethyl(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetate
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 6884761
Formula Weight 390.3g/mol
Chemical Name or Material 1,2-Diaminocyclohexanetetraacetic acid disodium salt solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.