Learn More
Medchemexpress LLC 1,2-dibenzoyl-1-(tert-butyl)hydrazine | 112225-87-3 | MFCD00940249 | 99.4% | 296.36 | C18H20N2O2 | 250 MG
SDP
Catalog No. 5000308273
Encompass_Preferred
RH-5849 (CAS 112225-87-3) is an orally active, non-steroidal ecdysteroid agonist used as a research insecticide reference. It inhibits larval growth of Plodia interpunctella in a dose-dependent manner and exhibits molting hormone activity. Supplied as a high-purity solid for laboratory research, typical specifications include chemical formula C18H20N2O2 and molecular weight 296.36.
Catalog No. 50-003-08273 Supplier Medchemexpress LLC Supplier No. HY118541250MG
RH-5849 (CAS 112225-87-3) is an orally active, non-steroidal ecdysteroid agonist used as a research insecticide reference. It inhibits larval growth of Plodia interpunctella in a dose-dependent manner and exhibits molting hormone activity. Supplied as a high-purity solid for laboratory research, typical specifications include chemical formula C18H20N2O2 and molecular weight 296.36.