Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibenzoylbenzene, 97%

Catalog No. AAL0159203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0159203 1 g
AAL01592MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0159203 Supplier Thermo Scientific Chemicals Supplier No. L0159203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1159-86-0 | C20H14O2 | 286.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159-86-0
Molecular Formula C20H14O2
Molecular Weight (g/mol) 286.33
MDL Number MFCD00003078
InChI Key OJLABXSUFRIXFL-UHFFFAOYSA-N
Synonym 1,2-dibenzoylbenzene, 1,2-phenylenebis phenylmethanone, 2-benzoylbenzophenone, 2-benzoylphenyl phenyl methanone, methanone, 1,2-phenylenebis phenyl, ortho-dibenzoylbenzene, o-dibenzoylbenzene, methanone,1,1'-1,2-phenylene bis 1-phenyl, phenyl 2-phenylcarbonyl phenyl ketone
PubChem CID 70875
IUPAC Name (2-benzoylphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2C(=O)C3=CC=CC=C3

Specifications

Melting Point 145°C to 148°C
Quantity 1 g
Beilstein 789077
Formula Weight 286.33
Percent Purity 97%
Chemical Name or Material 1,2-Dibenzoylbenzene
EINECSNumber 214-597-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.