Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibenzoylethylene, predominantly trans, 96%

Catalog No. AAL1264303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1264303 1 g
AAL1264306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1264303 Supplier Thermo Scientific Chemicals Supplier No. L1264303
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 959-28-4 | C16H12O2 | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-28-4
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00003083
InChI Key WYCXGQSQHAXLPK-VAWYXSNFSA-N
Synonym trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
PubChem CID 735960
IUPAC Name (E)-1,4-diphenylbut-2-ene-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 103°C to 108°C
Quantity 1 g
Beilstein 743958
Formula Weight 236.27
Percent Purity 96%
Chemical Name or Material 1,2-Dibenzoylethylene, predominantly trans
EINECSNumber 213-498-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.