Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-1,1,2,2-tetrachloroethane 97.0+%, TCI America™
SDP

Catalog No. D198725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D198725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D198725G Supplier TCI America Supplier No. D198725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 630-25-1
Molecular Formula C2Br2Cl4
Molecular Weight (g/mol) 325.63
MDL Number MFCD00000780
InChI Key WJUKOGPNGRUXMG-UHFFFAOYSA-N
Synonym 1,2-dibromotetrachloroethane, dbtce, sym-dibromotetrachloroethane, ethane, 1,2-dibromo-1,1,2,2-tetrachloro, unii-0sgg217efr, 0sgg217efr, 1,2-dibrom-1,1,2,2-tetrachlorethan, brccl2ccl2br, 1,2-dibromotetrachloroethan, 1,2-dibrom-tetrachlorethane
PubChem CID 69426
IUPAC Name 1,2-dibromo-1,1,2,2-tetrachloroethane
SMILES C(C(Cl)(Cl)Br)(Cl)(Cl)Br

Specifications

Color White
Quantity 25 g
Formula Weight 325.63
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dibromo-1,1,2,2-tetrachloroethane
RTECSNumber KH9366000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.