Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-2-methylpropane, 98%

Catalog No. p-102231
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406490050 5 g Glass bottle
AC406490250 25 g Glass bottle
AC406492500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC406490050 Supplier Thermo Scientific Chemicals Supplier No. 406490050
Only null left
Add to Cart

CAS: 594-34-3 | C4H8Br2 | 215.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-34-3
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.92
MDL Number MFCD00000126
InChI Key SDTXSEXYPROZSZ-UHFFFAOYSA-N
Synonym isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane
PubChem CID 222657
IUPAC Name 1,2-dibromo-2-methylpropane
SMILES CC(C)(CBr)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.7900g/mL
Boiling Point 150.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5083 to 1.5103
Linear Formula (CH3)2CBrCH2Br
Quantity 5 g
Specific Gravity 1.79
Solubility Information Solubility in water: slightly soluble
Formula Weight 215.92
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Dibromo-2-methylpropane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-835-7
RTECSNumber TX8810000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.