Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromoethane-d{4}, 99%(Isotopic)

Catalog No. AA4227906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4227906 5 g
AA4227914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4227906 Supplier Thermo Scientific Chemicals Supplier No. 04227906
Add to Cart

CAS: 22581-63-1 | C2H4Br2 | 191.886 g/mol

It is employed as a mutagenesis research chemical. Dibromoethane was shown to cause DNA single-strand breaks in isolated rat hepatocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22581-63-1
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 191.886
MDL Number MFCD00037499
InChI Key PAAZPARNPHGIKF-LNLMKGTHSA-N
Synonym 1,2-dibromoethane-d4, 1,2-dibromo-1,1,2,2-2h4 ethane, dibromo 2 h? ethane, dibromoethane-d4, ethane-d4, 1,2-dibromo, ethylene-d4 dibromide, 2h4-edb, tetradeutero-1,2-dibromoethane, 1,2-dibromoethane-d4, 99 atom % d, 1,2-dibromoethane-d4 99.0 atom % d
PubChem CID 123404
IUPAC Name 1,2-dibromo-1,1,2,2-tetradeuterioethane
SMILES C(CBr)Br

Specifications

Physical Form Liquid
Quantity 5 g
Odor Slightly Sweetish
Density 2.20 g/mL
Refractive Index 1.536
Melting Point 9°C
Boiling Point 132°C
UN Number UN1605
Solubility Information Insoluble in water. Soluble in chloroform.
Formula Weight 191.89
Percent Purity 99%
Chemical Name or Material 1,2-Dibromoethane-d4, Isotopic
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335-H350
Hazard Statement GHS H Statement
H301-H311-H331-H350-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ETHYLENE DIBROMIDE
EINECSNumber 245-102-8
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.