Learn More
Medchemexpress LLC 1,2-dibromoethane-d4 | 22581-63-1 | MFCD00037499 | >98.0% | 191.89 g·mol⁻¹ | C2Br2D4 | 100 MG
SDP
Catalog No. 5000284295
Encompass_Preferred
1,2-dibromoethane-d4 is the deuterium-labeled analogue of ethylene dibromide supplied as a neat liquid for use as an isotopic reagent, internal standard, or NMR solvent in research and synthetic applications. The product has formula C2Br2D4, CAS 22581-63-1, and a molecular weight of approximately 191.89 g·mol⁻¹. It is provided with high isotopic enrichment and chemical purity for analytical reliability. For research use only, not for human consumption.
  • Deuterium-labeled (d4) for isotopic tracing and NMR applications.
  • High isotopic enrichment suitable for quantitative analysis.
  • Chemical purity typically >98.0% to ensure analytical reliability.
  • Useful as an internal standard and reagent in synthesis and spectroscopy.
  • Liquid form with formula C2Br2D4 and molecular weight 191.89 g·mol⁻¹.
  • Store cold (refrigerated or -20°C recommended) to preserve stability.
Catalog No. 50-002-84295 Supplier Medchemexpress LLC Supplier No. HYW578702S100MG
1,2-dibromoethane-d4 is the deuterium-labeled analogue of ethylene dibromide supplied as a neat liquid for use as an isotopic reagent, internal standard, or NMR solvent in research and synthetic applications. The product has formula C2Br2D4, CAS 22581-63-1, and a molecular weight of approximately 191.89 g·mol⁻¹. It is provided with high isotopic enrichment and chemical purity for analytical reliability. For research use only, not for human consumption.
  • Deuterium-labeled (d4) for isotopic tracing and NMR applications.
  • High isotopic enrichment suitable for quantitative analysis.
  • Chemical purity typically >98.0% to ensure analytical reliability.
  • Useful as an internal standard and reagent in synthesis and spectroscopy.
  • Liquid form with formula C2Br2D4 and molecular weight 191.89 g·mol⁻¹.
  • Store cold (refrigerated or -20°C recommended) to preserve stability.