Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dibromoethane-d4 | 22581-63-1 | MFCD00037499 | >99.0% | 191.88 | C2H4Br2D4 | 500 MG
SDP

Catalog No. 5000284294
Encompass_Preferred

1,2-Dibromoethane-d4 is a deuterium-labeled analog of 1,2-dibromoethane used as an isotopically labeled reagent and NMR solvent in research and analytical workflows.

  • High isotopic enrichment (≥99.5%).
  • Chemical purity >99.0%.
  • Suitable for NMR spectroscopy and synthetic applications.
  • Provides known chemical identifiers: CAS 22581-63-1, formula C2H4Br2D4, MW 191.88.
  • Available as a 500 mg quantity for laboratory use.

Catalog No. 50-002-84294 Supplier Medchemexpress LLC Supplier No. HYW578702S500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dibromoethane-d4 is a deuterium-labeled analog of 1,2-dibromoethane used as an isotopically labeled reagent and NMR solvent in research and analytical workflows.

  • High isotopic enrichment (≥99.5%).
  • Chemical purity >99.0%.
  • Suitable for NMR spectroscopy and synthetic applications.
  • Provides known chemical identifiers: CAS 22581-63-1, formula C2H4Br2D4, MW 191.88.
  • Available as a 500 mg quantity for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.