Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dibromoethane-d4 | 22581-63-1 | MFCD00037499 | 99.0% | 191.89 g/mol | C2D4Br2 | 1 G
SDP

Catalog No. 5000284452
Encompass_Preferred

1,2-dibromoethane-d4 is the deuterium-labeled (d4) isotopologue of 1,2-dibromoethane used as an analytical standard and solvent in research and spectroscopic applications. It provides high isotopic enrichment and chemical purity suitable for NMR, tracer, and synthesis work.

  • Deuterium-labeled (d4) isotopologue of 1,2-dibromoethane.
  • Isotopic purity ~99 atom % D.
  • Assay purity 99.0%.
  • Molecular formula C2D4Br2; molecular weight 191.89 g/mol.
  • Liquid at ambient temperature; supplied in 1 G packages.
  • Intended for research and analytical applications.

Catalog No. 50-002-84452 Supplier Medchemexpress LLC Supplier No. HYW578702S1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-dibromoethane-d4 is the deuterium-labeled (d4) isotopologue of 1,2-dibromoethane used as an analytical standard and solvent in research and spectroscopic applications. It provides high isotopic enrichment and chemical purity suitable for NMR, tracer, and synthesis work.

  • Deuterium-labeled (d4) isotopologue of 1,2-dibromoethane.
  • Isotopic purity ~99 atom % D.
  • Assay purity 99.0%.
  • Molecular formula C2D4Br2; molecular weight 191.89 g/mol.
  • Liquid at ambient temperature; supplied in 1 G packages.
  • Intended for research and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.