Learn More
Medchemexpress LLC 1,2-dibromoethane-d4 | 22581-63-1 | MFCD00037499 | 99.0% | 191.89 g/mol | C2D4Br2 | 10 G
SDP
Catalog No. 5000284112
Encompass_Preferred
1,2-Dibromoethane-d4 is a deuterium-labeled analog of 1,2-dibromoethane used as an isotopically enriched reagent and reference standard in analytical and synthetic chemistry. It is commonly used in NMR studies and isotopic tracing applications and is provided for research use only.
  • Deuterium labeled (d4) for isotopic tracing and NMR studies.
  • High isotopic enrichment: 99.0% D.
  • Suitable as an NMR solvent or reference standard.
  • Useful as an isotopically labeled reagent in synthesis.
  • Molecular formula C2D4Br2; molecular weight 191.89 g/mol.
  • Supplied as 10 G quantity for laboratory use.
Catalog No. 50-002-84112 Supplier Medchemexpress LLC Supplier No. HYW578702S10G
1,2-Dibromoethane-d4 is a deuterium-labeled analog of 1,2-dibromoethane used as an isotopically enriched reagent and reference standard in analytical and synthetic chemistry. It is commonly used in NMR studies and isotopic tracing applications and is provided for research use only.
  • Deuterium labeled (d4) for isotopic tracing and NMR studies.
  • High isotopic enrichment: 99.0% D.
  • Suitable as an NMR solvent or reference standard.
  • Useful as an isotopically labeled reagent in synthesis.
  • Molecular formula C2D4Br2; molecular weight 191.89 g/mol.
  • Supplied as 10 G quantity for laboratory use.