Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-dibromoethane-d4 | 22581-63-1 | MFCD00067676 | >99.0% | 191.88 g/mol | C2D4Br2 | 250 MG
SDP

Catalog No. 5000283880
Encompass_Preferred

1,2-Dibromoethane-d4 is the deuterium-labeled isotopologue of 1,2-dibromoethane used as an analytical standard and reagent. It provides a mass-shifted analog for mass spectrometry, tracer studies, and synthetic applications where deuterium labeling is required.

  • High isotopic purity suitable for analytical use.
  • Molecular formula C2D4Br2 and molecular weight 191.88 g/mol.
  • CAS number 22581-63-1 for unambiguous identification.
  • Supplied in small, laboratory-scale quantity (250 mg).
  • Commonly used as an internal standard for GC-MS and LC-MS.
  • Useful for tracer studies and deuterium-labeling in synthesis.
  • Handle as a hazardous laboratory chemical and follow SDS recommendations.

Catalog No. 50-002-83880 Supplier Medchemexpress LLC Supplier No. HYW578702S250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2-Dibromoethane-d4 is the deuterium-labeled isotopologue of 1,2-dibromoethane used as an analytical standard and reagent. It provides a mass-shifted analog for mass spectrometry, tracer studies, and synthetic applications where deuterium labeling is required.

  • High isotopic purity suitable for analytical use.
  • Molecular formula C2D4Br2 and molecular weight 191.88 g/mol.
  • CAS number 22581-63-1 for unambiguous identification.
  • Supplied in small, laboratory-scale quantity (250 mg).
  • Commonly used as an internal standard for GC-MS and LC-MS.
  • Useful for tracer studies and deuterium-labeling in synthesis.
  • Handle as a hazardous laboratory chemical and follow SDS recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.