Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1,2-Dibromoethyl)benzene, 97%

Catalog No. AC299305000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299305000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC299305000 Supplier Thermo Scientific Chemicals Supplier No. 299305000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 93-52-7 | C8H8Br2 | 263.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-52-7
Molecular Formula C8H8Br2
Molecular Weight (g/mol) 263.95
MDL Number MFCD00000138
InChI Key SHKKTLSDGJRCTR-UHFFFAOYSA-N
Synonym 1,2-dibromoethyl benzene, dowspray 9, 1,2-dibromo-1-phenylethane, 1,2-dibromo phenyl ethane, benzene, 1,2-dibromoethyl, 1,2-dibromo-2-phenylethane, vic-styrene dibromide, alpha,beta-dibromoethylbenzene, ccris 2090, .alpha.,.beta.-dibromoethylbenzene
PubChem CID 7145
IUPAC Name 1,2-dibromoethylbenzene
SMILES C1=CC=C(C=C1)C(CBr)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 68.0°C to 73.0°C
Color Brown to Yellow
Density 1.74g/mL
Boiling Point 139.0°C to 141.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 05, 356
Formula Weight 263.95
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (1, 2-Dibromoethyl)benzene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-253-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.