Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromopropane Solution, 10,000μg/mL in hexane, Ultra Scientific

Catalog No. USPPS4001
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPPS4001 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPPS4001 Supplier Agilent Technologies Supplier No. PPS4001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-75-1 , 110-54-3
Molecular Formula C3H6Br2
Molecular Weight (g/mol) 201.889
InChI Key XFNJYAKDBJUJAJ-UHFFFAOYSA-N
PubChem CID 6553
IUPAC Name 1,2-dibromopropane
SMILES CC(CBr)Br

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 10, 000μg/mL
Chemical Name or Material 1, 2-Dibromopropane Solution, 10, 000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.