Learn More
Medchemexpress LLC 1,2-dichloro-3-methylbenzene-d3 | 1219798-57-8 | 164.05 g/mol | C7H3D3Cl2 | 100 MG

Supplier: Medchemexpress LLC HYW015427S100MG
1,2-Dichloro-3-methylbenzene-d3 is the deuterium-labeled isotopologue of 1,2-dichloro-3-methylbenzene used as an isotopic tracer in analytical chemistry and drug-development research. It is intended for quantitative mass spectrometry, metabolic profiling, and tracer studies, and is supplied for research use only.
- Deuterium-labeled (d3) isotopologue for tracer applications.
- Molecular formula C7H3D3Cl2; molecular weight 164.05 g/mol.
- Suitable for mass spectrometry quantitation and metabolic profiling.
- Listed as supplied in pure form on manufacturer documentation.
- Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
- For research use only; not validated for clinical or medical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.