Learn More
Medchemexpress LLC 1,2-dichloro-3-methylbenzene-d3 | 1219798-57-8 | 164.05 g/mol | C7H3D3Cl2 | 250 MG
SDP
Catalog No. 5000302840
Encompass_Preferred
1,2-Dichloro-3-methylbenzene-d3 is the deuterium-labeled analogue of 1,2-dichloro-3-methylbenzene used as an isotopic standard in analytical and research applications. It is intended for use in mass spectrometry quantitation, metabolic tracing, and analytical calibration, provided as a high-purity reagent for laboratory workflows.
  • Deuterium-labeled reference standard for mass spectrometry
  • High chemical and isotopic purity suitable for quantitation
  • Useful for metabolic tracing and internal standardization
  • Stable under recommended storage conditions for reliable performance
  • Supplied in a convenient small-quantity vial for laboratory use
Catalog No. 50-003-02840 Supplier Medchemexpress LLC Supplier No. 250MGHYW015427S
1,2-Dichloro-3-methylbenzene-d3 is the deuterium-labeled analogue of 1,2-dichloro-3-methylbenzene used as an isotopic standard in analytical and research applications. It is intended for use in mass spectrometry quantitation, metabolic tracing, and analytical calibration, provided as a high-purity reagent for laboratory workflows.
  • Deuterium-labeled reference standard for mass spectrometry
  • High chemical and isotopic purity suitable for quantitation
  • Useful for metabolic tracing and internal standardization
  • Stable under recommended storage conditions for reliable performance
  • Supplied in a convenient small-quantity vial for laboratory use