Learn More
Medchemexpress LLC 1,2-dichloro-3-methylbenzene-d3 | 1219798-57-8 | >98.0% | 164.05 g/mol | C7H3D3Cl2 | 50 MG

Supplier: Medchemexpress LLC HYW015427S50MG
1,2-dichloro-3-methylbenzene-d3 is the deuterium-labeled analogue of 1,2-dichloro-3-methylbenzene used as an isotope-labeled reference standard and tracer in analytical and drug metabolism studies. It contains three deuterium atoms on the methyl group and has molecular formula C7H3D3Cl2 and molecular weight 164.05 g/mol.
- Deuterium-labeled (d3) on the methyl group.
- Chemical formula C7H3D3Cl2.
- Molecular weight 164.05 g/mol.
- Used as an isotope-labeled reference standard and tracer in analytical and metabolism studies.
- Storage: pure form -20°C for up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
- Deuterium enrichment ≥98 atom % D.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.