Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,2-Dichlorobenzene solution NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm x 8 in. | 95-50-1 | MFCD00000535 | 10X8IN
Catalog No. 501860496
Encompass
1,2-Dichlorobenzene solution NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm x 8 in. | Mol Wt: 147 | 95-50-1 | MFCD00000535 | 10X8IN
This item is not returnable. View return policy
Catalog No. 50-186-0496 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 61250210X8IN
1,2-Dichlorobenzene solution NMR reference standard, 1% in acetone-d6 (99.9 atom % D), NMR tube size 3 mm x 8 in. | Mol Wt: 147 | 95-50-1 | MFCD00000535 | 10X8IN