Learn More
1,2-Dichloroethane, 99.8+%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Orange to Yellow |
| Density | 1.2500g/mL |
| Quantity | 100 mL |
| Infrared Spectrum | Authentic |
| Physical Form | Crystalline Powder |
| Refractive Index | 1.4440 to 1.4460 (20°C, 589nm) |
| Residue after Evaporation | 0.005% max. |
| Assay Percent Range | 99.8+% |
| Vapor Pressure | Vapor Pressure: 65mmHg at 29°C |
| Linear Formula | ClCH2CH2Cl |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause cancer. Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. Highly flammable liquid and vapor. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: IF exposed or concerned: Get medical advice/attention. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 203-458-1 |
| RTECSNumber | KI0525000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.